missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl 4-hydroxybenzoate, 98%

CAS: 4247-02-3 | C11H14O3 | 194.23 g/mol

Supplier:  Thermo Scientific Chemicals L1758718

 View more versions of this product

Catalog No. AAL1758718

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Isobutyl 4-hydroxybenzoate is used as an anti-microbial agent. It is also used as antiseptics for cosmetic field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

Isobutyl 4-hydroxybenzoate
74°C to 78°C
MFCD00020167
2642305
Insoluble in water.
CC(C)COC(=O)C1=CC=C(C=C1)O
194.23
194.23
2-3-4247
C11H14O3
50 g
isobutyl 4-hydroxybenzoate, isobutylparaben, isobutyl p-hydroxybenzoate, isobutyl paraben, isobutyl-4-hydroxybenzoate, isobutyl parahydroxybenzoate, 4-hydroxybenzoic acid isobutyl ester, benzoic acid, 4-hydroxy-, 2-methylpropyl ester, p-oxybenzoesaureisobutylester, unii-0qqj25x58g
XPJVKCRENWUEJH-UHFFFAOYSA-N
2-methylpropyl 4-hydroxybenzoate
20240
98%

Chemical Identifiers

2-3-4247
194.23
XPJVKCRENWUEJH-UHFFFAOYSA-N
20240
CC(C)COC(=O)C1=CC=C(C=C1)O
C11H14O3
MFCD00020167
isobutyl 4-hydroxybenzoate, isobutylparaben, isobutyl p-hydroxybenzoate, isobutyl paraben, isobutyl-4-hydroxybenzoate, isobutyl parahydroxybenzoate, 4-hydroxybenzoic acid isobutyl ester, benzoic acid, 4-hydroxy-, 2-methylpropyl ester, p-oxybenzoesaureisobutylester, unii-0qqj25x58g
2-methylpropyl 4-hydroxybenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-208-8

RTECSNumber : DH2247000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only