missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutyl 4-hydroxybenzoate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Isobutyl 4-hydroxybenzoate is used as an anti-microbial agent. It is also used as antiseptics for cosmetic field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Isobutyl 4-hydroxybenzoate is used as an anti-microbial agent. It is also used as antiseptics for cosmetic field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 74°C to 78°C |
| Quantity | 5 g |
| Beilstein | 2642305 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 194.23 |
| Percent Purity | 98% |
| Chemical Name or Material | Isobutyl 4-hydroxybenzoate |
Safety and Handling
| EINECSNumber | 224-208-8 |
| RTECSNumber | DH2247000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only