missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobornyl Acetate 90.0+%, TCI America™
$49.36 - $96.84
Chemical Identifiers
| CAS | 125-12-2 |
|---|---|
| Molecular Formula | C12H20O2 |
| Molecular Weight (g/mol) | 196.29 |
| MDL Number | MFCD00867808,MFCD00135943 |
| InChI Key | KGEKLUUHTZCSIP-JFGNBEQYSA-N |
| Synonym | 1s,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl acetate |
| PubChem CID | 6950273 |
| IUPAC Name | (1S,2S,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate |
| SMILES | CC(=O)O[C@H]1C[C@H]2CC[C@@]1(C)C2(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
I030625ML
|
N/A | 25 mL |
Each for $49.36
|
|
|||||
|
I0306500ML
|
N/A | 500 mL |
Each for $96.84
|
|
|||||
Chemical Identifiers
| C12H20O2 | |
| MFCD00867808,MFCD00135943 | |
| 1s,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl acetate | |
| (1S,2S,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate |
Specifications
| 125-12-2 | |
| Yellow | |
| C12H20O2 | |
| 1s,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl acetate | |
| CC(=O)O[C@H]1C[C@H]2CC[C@@]1(C)C2(C)C | |
| 196.29 | |
| 196.29 | |
| Liquid |
| 7°C | |
| 106°C | |
| MFCD00867808,MFCD00135943 | |
| KGEKLUUHTZCSIP-JFGNBEQYSA-N | |
| (1S,2S,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate | |
| 6950273 | |
| ≥90.0% (GC) | |
| Isobornyl Acetate |
Safety and Handling
EINECSNumber : (4)-0626&(4)-1216
RTECSNumber : NP7350000
TSCA : Yes