accessibility menu, dialog, popup

UserName

H3COO(S)(R)(S)CH3H3CH3C

CAS RN 125-12-2

IUPAC Name: (1S,2S,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
Synonyms: isobornyl acetate
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: KGEKLUUHTZCSIP-JFGNBEQYSA-N
SMILES: CC(=O)O[C@H]1C[C@H]2CC[C@@]1(C)C2(C)C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)