missing translation for 'onlineSavingsMsg'
Learn More

Isobornyl Acetate 90.0+%, TCI America™

Catalog No. I030625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I030625ML 25 mL
I0306500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I030625ML Supplier TCI America Supplier No. I030625ML

Chemical Identifiers

CAS 125-12-2
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135943
InChI Key KGEKLUUHTZCSIP-JFGNBEQYSA-N
Synonym 1s,4r-1,7,7-trimethylbicyclo 2.2.1 heptan-2-yl acetate
PubChem CID 6950273
IUPAC Name (1S,2S,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate
SMILES CC(=O)O[C@H]1C[C@H]2CC[C@@]1(C)C2(C)C

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 106°C
Quantity 25 mL
Formula Weight 196.29
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobornyl Acetate
EINECSNumber (4)-0626&(4)-1216
RTECSNumber NP7350000
TSCA Yes