missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Methyl Ketone 98.0+%, TCI America™
Supplier: TCI America I0087500ML
Specifications
| Isoamyl Methyl Ketone | |
| -74°C | |
| 144°C | |
| MFCD00008950 | |
| 2302 | |
| FFWSICBKRCICMR-UHFFFAOYSA-N | |
| 5-methylhexan-2-one | |
| 8034 | |
| ≥98.0% (GC) |
| 110-12-3 | |
| Colorless | |
| C7H14O | |
| 500 mL | |
| 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28 | |
| CC(C)CCC(=O)C | |
| 114.188 | |
| 114.19 | |
| Liquid |
Chemical Identifiers
| 110-12-3 | |
| 114.188 | |
| FFWSICBKRCICMR-UHFFFAOYSA-N | |
| 8034 | |
| CC(C)CCC(=O)C |
| C7H14O | |
| MFCD00008950 | |
| 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28 | |
| 5-methylhexan-2-one |
Safety and Handling
EINECSNumber : (2)-0542
RTECSNumber : MP3850000
TSCA : Yes