accessibility menu, dialog, popup

UserName

H3CCH3CH3O

CAS RN 110-12-3

IUPAC Name: 5-methylhexan-2-one
Synonyms: 5-methyl-2-hexanone
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: FFWSICBKRCICMR-UHFFFAOYSA-N
SMILES: CC(C)CCC(C)=O
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)