missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl Methyl Ketone 98.0+%, TCI America™

Catalog No. p-7131205
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
I008725ML 25 mL
I0087500ML 500 mL
2 options

Catalog No. I008725ML

Supplier: TCI America I008725ML

Only null left

Chemical Identifiers

CAS 110-12-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008950
InChI Key FFWSICBKRCICMR-UHFFFAOYSA-N
Synonym 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
PubChem CID 8034
IUPAC Name 5-methylhexan-2-one
SMILES CC(C)CCC(=O)C

Specifications

Melting Point -74°C
Color Colorless
Boiling Point 144°C
Quantity 25 mL
UN Number 2302
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl Methyl Ketone
EINECSNumber (2)-0542
RTECSNumber MP3850000
TSCA Yes