missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoamyl Benzoate 98.0+%, TCI America™
$46.60 - $203.17
Chemical Identifiers
| CAS | 94-46-2 |
|---|---|
| Molecular Formula | C12H16O2 |
| Molecular Weight (g/mol) | 192.258 |
| MDL Number | MFCD00026515 |
| InChI Key | MLLAPOCBLWUFAP-UHFFFAOYSA-N |
| Synonym | Benzoic Acid Isoamyl Ester, Isopentyl Benzoate |
| PubChem CID | 7193 |
| IUPAC Name | 3-methylbutyl benzoate |
| SMILES | CC(C)CCOC(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 94-46-2 | |
| 192.258 | |
| MLLAPOCBLWUFAP-UHFFFAOYSA-N | |
| 7193 | |
| CC(C)CCOC(=O)C1=CC=CC=C1 |
| C12H16O2 | |
| MFCD00026515 | |
| Benzoic Acid Isoamyl Ester, Isopentyl Benzoate | |
| 3-methylbutyl benzoate |
Specifications
| 94-46-2 | |
| 262°C | |
| MFCD00026515 | |
| Benzoic Acid Isoamyl Ester, Isopentyl Benzoate | |
| CC(C)CCOC(=O)C1=CC=CC=C1 | |
| 192.258 | |
| 192.26 | |
| Liquid |
| Colorless | |
| C12H16O2 | |
| 25 g | |
| MLLAPOCBLWUFAP-UHFFFAOYSA-N | |
| 3-methylbutyl benzoate | |
| 7193 | |
| ≥98.0% (GC) | |
| Isoamyl Benzoate |
Safety and Handling
EINECSNumber : (3)-1356
RTECSNumber : DH3078000
TSCA : Yes