missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl 4-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. h021410g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
Catalog No. Quantity
H021410G 10 g
H021425G 25 g
2 options

Catalog No. H021410G

Supplier: TCI America H021410G

Only null left

Chemical Identifiers

CAS 6521-30-8
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00017502
InChI Key KSHVDKDQYBNSAN-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid Isoamyl Ester, Isoamylparaben, Isopentyl 4-Hydroxybenzoate
PubChem CID 81009
IUPAC Name 3-methylbutyl 4-hydroxybenzoate
SMILES CC(C)CCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 62°C
Color White
Boiling Point 242°C
Quantity 10 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isoamyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
TSCA Yes