missing translation for 'onlineSavingsMsg'
Learn More

Iso-Propyl Alcohol, 70% v/v, Reagents

Catalog No. 01334905
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334905 20 L
01334906 1 L
01334904 4 L
01334903 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01334905 Supplier Labchem Inc Supplier No. CI29600020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iso-Propyl Alcohol, 70% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropyl Alcohol, 2-Propanol, Propan-2-ol
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Quantity 20 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 0.877
Boiling Point 88°C
Percent Purity 70%
Grade Reagent
Chemical Name or Material Iso-Propyl Alcohol, 70% v/v