missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Octane, PRF, B&J Brand, meets requirements of ASTM D2699 and D2700, Honeywell Burdick & Jackson
$7530.43 - $13069.78
Chemical Identifiers
| CAS | 540-84-1 |
|---|---|
| Molecular Formula | C8H18 |
| Molecular Weight (g/mol) | 114.232 |
| InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
| Synonym | isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane |
| PubChem CID | 10907 |
| ChEBI | CHEBI:62805 |
| IUPAC Name | 2,2,4-trimethylpentane |
| SMILES | CC(C)CC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6000875
|
Honeywell-Burdick and Jackson
RS36219 |
19 L | Steel Can |
Each for $7,530.43
|
|
||||
|
6000876
|
Honeywell-Burdick and Jackson
RS362204 |
204 L | Steel Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 540-84-1 | |
| 114.232 | |
| isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
| CHEBI:62805 | |
| CC(C)CC(C)(C)C |
| C8H18 | |
| NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
| 10907 | |
| 2,2,4-trimethylpentane |
Specifications
| 540-84-1 | |
| 0.69 g/cm3 | |
| 19 L | |
| Steel Can | |
| UN1262 | |
| NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
| 2,2,4-trimethylpentane | |
| 10907 | |
| 114.23 | |
| Primary Reference Fuel |
| 100.0000% | |
| 99.24°C | |
| −8°C | |
| C8H18 | |
| isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
| CC(C)CC(C)(C)C | |
| 114.232 | |
| CHEBI:62805 | |
| >99.75% | |
| Iso-Octane, PRF, B&J Brand, meets requirements of ASTM D2699 and D2700 |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes