missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Octane, PRF, B&J Brand, meets requirements of ASTM D2699 and D2700, Honeywell Burdick & Jackson
Specifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.69 g/cm3 |
| Quantity | 204 L |
| Flash Point | −8°C |
| Packaging | Steel Drum |
| UN Number | UN1262 |
| Boiling Point | 99.24°C |
| Formula Weight | 114.23 |
| Percent Purity | >99.75% |
| Grade | Primary Reference Fuel |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes |