missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol), Reagents
Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C11203004C
Specifications
| Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol) | |
| 78-83-1 | |
| 108°C | |
| Glass Bottle | |
| 0.8016 | |
| Isobutanol, 2-Methyl-1-propanol, 2-Methylpropan-1-ol | |
| CC(C)CO | |
| 74.12 |
| -108°C | |
| Colorless | |
| 4 L | |
| Liquid | |
| C4H10O | |
| ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
| 2-methylpropan-1-ol | |
| ACS |
Chemical Identifiers
| 78-83-1 | |
| 74.12 | |
| Isobutanol, 2-Methyl-1-propanol, 2-Methylpropan-1-ol | |
| CC(C)CO |
| C4H10O | |
| ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
| 2-methylpropan-1-ol |