missing translation for 'onlineSavingsMsg'
Learn More

Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol), Reagents

Catalog No. p-200020994
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338150 4 L Poly Bottle
01338149 4 L Glass Bottle
01338148 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338150 Supplier Labchem Inc Supplier No. C11203004A
Only null left

Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym Isobutanol, 2-Methyl-1-propanol, 2-Methylpropan-1-ol
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point -108°C
Color Colorless
Boiling Point 108°C
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.8016
Grade ACS
Chemical Name or Material Iso-Butyl Alcohol, ACS Reagent (2-Methyl-1-Propanol)