missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron Standard, 200 ppm Fe, Ricca Chemical
$123.87 - $123.87
Chemical Identifiers
| CAS | 7783-83-7 |
|---|---|
| Molecular Formula | FeH28NO20S2 |
| Molecular Weight (g/mol) | 482.176 |
| InChI Key | LCPUDZUWZDSKMX-UHFFFAOYSA-K |
| PubChem CID | 197097 |
| IUPAC Name | azanium;iron(3+);disulfate;dodecahydrate |
| SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Chemical Identifiers
| 7783-83-7 | |
| 482.176 | |
| 197097 | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| azanium;iron(3+);disulfate;dodecahydrate |
Specifications
| Amber Glass Bottle | |
| 97.0,0.45,0.15 | |
| 100.0,0.46,0.16 | |
| FeH28NO20S2 | |
| 100°C | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
| 482.176 | |
| 197097 |
| 7783-83-7,7664-93-9,7732-18-5 | |
| 200ppm Fe | |
| Liquid | |
| Green/Yellow | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| azanium;iron(3+);disulfate;dodecahydrate | |
| 120 mL | |
| Iron Standard Stock Solution |