missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™
Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric)
Supplier: Honeywell-Fluka 123544X5KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Iron(Ii) Sulfate Heptahydrate | |
| 7782-63-0 | |
| FeH14O11S | |
| MFCD00149719 | |
| SURQXAFEQWPFPV-UHFFFAOYSA-L | |
| iron(2+);sulfate;heptahydrate | |
| 62662 | |
| 278.01 | |
| puriss. | |
| 1.898g/mL at 25°C |
| Meets analytical specification of Ph. Eur.BP, USP, FCC | |
| 4 x 5 kg | |
| FeSO4 · 7H2O | |
| iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
| O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O | |
| 278.01 | |
| CHEBI:75836 | |
| 99.5 to 104.5% (manganometric) | |
| 14.6mmHg (25°C) |
Chemical Identifiers
| 7782-63-0 | |
| 278.01 | |
| SURQXAFEQWPFPV-UHFFFAOYSA-L | |
| 62662 | |
| iron(2+);sulfate;heptahydrate |
| FeH14O11S | |
| MFCD00149719 | |
| iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
| CHEBI:75836 | |
| O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
Safety and Handling
P301 + P312 + P330-P305 + P351 + P338
H302-H315-H319
EINECSNumber : 231-753-5
RTECSNumber : NO8510000