missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™

Catalog No. p-7071783
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001934 1 kg
6001938 5 kg
60044731 6 x 1 kg
60044730 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001934 Supplier Honeywell Chemicals Supplier No. 123541KG
Only null left

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name iron(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula FeSO4 · 7H2O
Formula Weight 278.01g/mol
Percent Purity 99.5 to 104.5% (manganometric)
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 14.6mmHg (25°C)
Density 1.898g/mL at 25°C
Chemical Name or Material Iron(Ii) Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
Hazard Statement P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302-H315-H319
EINECSNumber 231-753-5
RTECSNumber NO8510000