missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Sulfate, Heptahydrate, ACS, 99.5 to 102.00%, MilliporeSigma™

Supplier:  Technidata 1039651000

 View more versions of this product

Catalog No. M1039651000


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Iron(II) Sulfate, Heptahydrate
Blue to Green or White
Crystals
99.5 to 102.00%
FeSO4 · 7H2O
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
278.01
CHEBI:75836
ACS/EP/ISO/Reagent
EMSURE™, For Analysis
7782-63-0
1 kg
FeH14O11S
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
λ²-iron(2+) heptahydrate sulfate
62662
278.2
1.89g/cm3 (20°C); Bulk Density: 600 kg/m3

Chemical Identifiers

7782-63-0
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
λ²-iron(2+) heptahydrate sulfate
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
CHEBI:75836
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at +15°C to 25°C

SDS
Documents

Documents

Promotions

Promotions