missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) Sulfate, Heptahydrate, ACS, 99.5 to 102.00%, MilliporeSigma™

Catalog No. p-4381938
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1039650500 500 g
M1039651000 1 kg
M1039655000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1039650500 Supplier Technidata Supplier No. 1.03965.0500
Only null left

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Color Blue-Green or White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99.5 to 102.00%
Linear Formula FeSO4 · 7H2O
Formula Weight 278.2
Grade ACS/EP/ISO/Reagent
Density 1.89g/cm3 (20°C); Bulk Density: 600 kg/m3
Chemical Name or Material Iron(II) Sulfate, Heptahydrate, EMSURE™, For Analysis
Recommended Storage Store at +15°C to 25°C