Learn More
Eisco™ Iron Hooked Weight
Hooks included on both sides can be removed for versatility and hanging in tandem. Weights are painted black with weight value painted in yellow.
$62.07 - $79.79
Specifications
| Product Type | Cylindrical Weight |
|---|---|
| Material | Iron |
| For Use With (Application) | Calibration, mass measurment, physics experiments |
| Includes | 1 weight, 2 hooks |
| Quantity | 1 |
Description
- Solid Iron Hooked Weight
- Retractable hooks included on both sides for linking weights in tandem
- Hooks connect to weight via bored hole, allowing weight to sit on a flat surface when hook is removed
- Weight is painted black with yellow weight value painted on top
Warranty
1 year limited manufacturer warranty
Specifications
| Cylindrical Weight | |
| Calibration, mass measurment, physics experiments | |
| 1 |
| Iron | |
| 1 weight, 2 hooks |