missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals IR-775 chloride, 90% dye content
CAS: 199444-11-6 | C32H36Cl2N2 | 519.55 g/mol
$164.18 - $466.04
Chemical Identifiers
| CAS | 199444-11-6 |
|---|---|
| Molecular Formula | C32H36Cl2N2 |
| Molecular Weight (g/mol) | 519.55 |
| MDL Number | MFCD03427067,MFCD03427067 |
| InChI Key | BPSIJFMUSNMMAL-UHFFFAOYSA-M |
| Synonym | ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride |
| PubChem CID | 71311114 |
| IUPAC Name | 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride |
| SMILES | [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH66770MD
|
Thermo Scientific Chemicals
H66770MD |
250 mg |
Each for $164.18
|
|
|||||
|
AAH6677003
|
Thermo Scientific Chemicals
H6677003 |
1 g |
Each for $466.04
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 199444-11-6 | |
| 519.55 | |
| BPSIJFMUSNMMAL-UHFFFAOYSA-M | |
| 71311114 | |
| [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl |
| C32H36Cl2N2 | |
| MFCD03427067,MFCD03427067 | |
| ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride | |
| 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride |
Specifications
| 199444-11-6 | |
| Green | |
| MFCD03427067,MFCD03427067 | |
| ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride | |
| [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl | |
| 519.55 | |
| 519.55 | |
| IR-775 chloride, 90% dye content |
| 205°C to 214°C | |
| C32H36Cl2N2 | |
| 250 mg | |
| BPSIJFMUSNMMAL-UHFFFAOYSA-M | |
| 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride | |
| 71311114 | |
| Solid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only