missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals IR-775 chloride, 90% dye content
CAS: 199444-11-6 | C32H36Cl2N2 | 519.55 g/mol
Supplier: Thermo Scientific Chemicals H66770MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| IR-775 chloride | |
| 199444-11-6 | |
| Green | |
| MFCD03427067,MFCD03427067 | |
| ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride | |
| [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl | |
| 519.55 | |
| 519.55 |
| 90% dye content | |
| 205°C to 214°C | |
| C32H36Cl2N2 | |
| 250 mg | |
| BPSIJFMUSNMMAL-UHFFFAOYSA-M | |
| 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride | |
| 71311114 | |
| Solid |
Chemical Identifiers
| 199444-11-6 | |
| 519.55 | |
| BPSIJFMUSNMMAL-UHFFFAOYSA-M | |
| 71311114 | |
| [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl |
| C32H36Cl2N2 | |
| MFCD03427067,MFCD03427067 | |
| ir-775 chloride, 2-2-2-chloro-3-2-1,3-dihydro-1,3,3-trimethyl-2h-indol-2-ylidene-ethylidene-1-cyclohexen-1-yl-ethenyl-1,3,3-trimethyl-3h-indolium chloride, 2-2-2-chloro-3-2-1,3,3-trimethyl-1,3-dihydro-2h-indol-2-ylidene ethylidene cyclohex-1-en-1-yl ethenyl-1,3,3-trimethyl-3h-indol-1-ium chloride | |
| 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only