missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ iQ Series™ Men's Comfort Knit Black Flame Resistant Henley
iQ Series™ Men's Comfort Knit Black Flame Resistant Henley is CAT 2 AR/FR, NFPA 2112 compliant and a casual, comfortable and stylish part of your FR system. With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better.
$191.55 - $203.50
Specifications
| Gender | Men's |
|---|---|
| Material | Cotton, Polyester, POD |
| Color | Black |
| Neck Style | Henley |
| Sleeve Style | Raglan Sleeve |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-204-005
|
Bulwark™
QT20BKRGS |
Small |
Each for $203.50
|
|
|||||
|
17-103-755
|
Bulwark™
QT20BKRGM |
Medium |
Each for $191.55
|
|
|||||
|
17-204-062
|
Bulwark™
QT20BKRGL |
Large |
Each for $203.50
|
|
|||||
|
17-204-347
|
Bulwark™
QT20BKRGXL |
X-Large |
Each for $203.50
|
|
|||||
|
17-203-944
|
Bulwark™
QT20BKRGXXL |
2X-Large |
Each for $203.50
|
|
|||||
|
17-204-246
|
Bulwark™
QT20BKRG3XL |
3X-Large |
Each for $203.50
|
|
|||||
|
17-204-287
|
Bulwark™
QT20BKRG4XL |
4X-Large |
Each for $203.50
|
|
|||||
|
17-204-162
|
Bulwark™
QT20BKLNL |
Large (Long) |
Each for $203.50
|
|
|||||
|
17-204-360
|
Bulwark™
QT20BKLNXL |
X-Large (Long) |
Each for $203.50
|
|
|||||
|
17-204-143
|
Bulwark™
QT20BKLNXXL |
2X-Large (Long) |
Each for $203.50
|
|
|||||
|
17-204-188
|
Bulwark™
QT20BKLN3XL |
3X-Large (Long) |
Each for $203.50
|
|
|||||
Description
- One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
- Hidden rib knit cuffs to keep sleeves in place
- Moisture-wicking, breathable fabric
- Four-button placket
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
- Blend: 69% Cotton/ 25% Polyester/ 6% Polyoxadiazole
- Packaging: Polybag
Specifications
| Men's | |
| Black | |
| Raglan Sleeve | |
| Button | |
| 2 | |
| ASTM F1506, NFPA 2112, NFPA 70E |
| Cotton, Polyester, POD | |
| Henley | |
| Knit | |
| Reusable | |
| Category 2 |