missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ iQ Series™ Men's Comfort Knit Black Flame Resistant Henley
Description
- One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
- Hidden rib knit cuffs to keep sleeves in place
- Moisture-wicking, breathable fabric
- Four-button placket
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
- Blend: 69% Cotton/ 25% Polyester/ 6% Polyoxadiazole
- Packaging: Polybag
Specifications
Specifications
| Size | 2X-Large (Long) |
| Gender | Men's |
| Material | Cotton, Polyester, POD |
| Color | Black |
| Neck Style | Henley |
| Sleeve Style | Raglan Sleeve |
| Construction | Knit |
| Closure Type | Button |
| Disposable | Reusable |
| No. of Pockets | 2 |
| Show More |