missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ iQ Series™ Men's Comfort Knit Black Flame Resistant Henley
iQ Series™ Men's Comfort Knit Black Flame Resistant Henley is CAT 2 AR/FR, NFPA 2112 compliant and a casual, comfortable and stylish part of your FR system. With raglan sleeves and underarm gussets for added mobility, so you can bend, reach, stretch and work better.
Supplier: Bulwark™ QT20BKRGL
Description
- One concealed chest pocket with pencil stall and one concealed pencil stall sleeve pocket
- Hidden rib knit cuffs to keep sleeves in place
- Moisture-wicking, breathable fabric
- Four-button placket
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Exclusively with Westex G2™ fabrics by Milliken™ - Flame resistant, 5.3 oz. (180 g/m2)
- Blend: 69% Cotton/ 25% Polyester/ 6% Polyoxadiazole
- Packaging: Polybag
Specifications
| Large | |
| Cotton, Polyester, POD | |
| Henley | |
| Knit | |
| Reusable | |
| Category 2 |
| Men's | |
| Black | |
| Raglan Sleeve | |
| Button | |
| 2 | |
| ASTM F1506, NFPA 2112, NFPA 70E |