missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IPTG, dioxane-free

Catalog No. FERR0393
Change view
Click to view available options
Quantity:
5 g
5 x 5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0393 5 x 5 g
FERR0392 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERR0393 Supplier Thermo Scientific™ Supplier No. R0393

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Induce the expression of genes under the control of lac operon in cloning and protein expression procedures.

IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose. It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon . It is used in conjunction with X-Gal (#R0941) to determine the lac phenotype in blue/white colony screening.

It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon. It is used in conjunction with X-Gal to determine the lac phenotype in blue/white colony screening.

Ready-to-use 100mM IPTG aqueous solution is non-toxic, stable, 0.22μm membrane-filtered, formulated for direct use in blue/white colony screening.

Specifications

Chemical Name or Material Isopropylthiogalactoside
Concentration 0.1 mM
Product Type IPTG Solution
Quantity 5 x 5 g
Form Powder

For Research Use Only. Not for use in diagnostic procedures.