missing translation for 'onlineSavingsMsg'
Learn More

IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™

Catalog No. a4594
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Poly Bottle
PolyPac™
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A4594 4 L Poly Bottle
A459500 500 mL Poly Bottle
A4591 1 L Poly Bottle
A45920 20 L PolyPac™
A459200 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A4594 Supplier Thermo Fisher Scientific Supplier No. FLA4594
Only null left

Chemical Identifiers

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -88°C
Color Colorless
Density 0.7850g/cm³
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Refractive Index 1.3706 to 1.3722 (at 25°C)
Assay Percent Range Assay: 68.0 to 72.0%
Vapor Pressure 33mmHg at 20°C
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Boiling Point 82°C
Molecular Weight (g/mol) 60.096
Formula Weight 60.1
Percent Purity 68.0 to 72.0%
Chemical Name or Material IPA (Isopropyl Alcohol), 70% v/v
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:1
Flammability:3
Instability:0