missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ionomycin, 96%

Highly selective and potent calcium ionophore | CAS: 56092-81-0 | C41H72O9, C41H72O9 | 709.02 g/mol

$200.44 - $721.23

Chemical Identifiers

CAS 56092-81-0
Molecular Formula C41H72O9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ62448MCR
View Documents Promotion Details
Thermo Scientific Chemicals
J62448MCR
1 mg
Each for $200.44
Only null left
 
AAJ62448LB0
View Documents Promotion Details
Thermo Scientific Chemicals
J62448LB0
5 mg
Each for $721.23
Only null left
 
Description

Description

Ionomycin free acid is a beneficial tool in research to raise intracellular calcium levels. Also this antibiotic can be used to study the calcium transport across the plasma membrane. It has been shown to induce central demyelination , inhibit adrenal bovine TREK-1 channels , and to regulate cell division of mature human B cells . It is used to study the effects of calcium flux on endoplasmic reticulum (ER) stress, mitochondrial stress and intrinsic apoptosis mechanisms. It is also used to stimulate the intracellular production of the cytokines, interferon, perforin, IL-2, and IL-4 usually in conjunction with PMA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56092-81-0
C41H72O9

Specifications

Colorless
Highly selective and potent calcium ionophore
C41H72O9
Alcohol-like
Solution in ethanol
PGHMRUGBZOYCAA-OJFQOPKISA-N
(4R,6S,8S,10Z,12R,14R,16E,18R,19R,20S,21S)-11,19,21-trihydroxy-22-[(2S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9-oxodocosa-10,16-dienoic acid
98%
-30°C to -10°C
56092-81-0
709.02
Ionomycin
3642126
C[C@H](CCC(O)=O)C[C@H](C)C[C@H](C)C(=O)\C=C(/O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)CC1CC[C@](C)(O1)C1CC[C@](C)(O1)[C@@H](C)O
709.02
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only