Learn More
Thermo Scientific Chemicals Ionomycin, 96%
Highly selective and potent calcium ionophore | CAS: 56092-81-0 | C41H72O9, C41H72O9 | 709.02 g/mol
Supplier: Thermo Scientific Chemicals J62448LB0
| Quantity | 5 mg |
|---|
Description
Ionomycin free acid is a beneficial tool in research to raise intracellular calcium levels. Also this antibiotic can be used to study the calcium transport across the plasma membrane. It has been shown to induce central demyelination , inhibit adrenal bovine TREK-1 channels , and to regulate cell division of mature human B cells . It is used to study the effects of calcium flux on endoplasmic reticulum (ER) stress, mitochondrial stress and intrinsic apoptosis mechanisms. It is also used to stimulate the intracellular production of the cytokines, interferon, perforin, IL-2, and IL-4 usually in conjunction with PMA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56092-81-0 | |
| C41H72O9 |
Specifications
| 5 mg | |
| -30°C to -10°C | |
| 98% | |
| C41H72O9 | |
| Alcohol-like | |
| Solution in ethanol | |
| PGHMRUGBZOYCAA-OJFQOPKISA-N | |
| (4R,6S,8S,10Z,12R,14R,16E,18R,19R,20S,21S)-11,19,21-trihydroxy-22-[(2S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9-oxodocosa-10,16-dienoic acid |
| Colorless | |
| Highly selective and potent calcium ionophore | |
| 56092-81-0 | |
| 709.02 | |
| Ionomycin | |
| 3642126 | |
| C[C@H](CCC(O)=O)C[C@H](C)C[C@H](C)C(=O)\C=C(/O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)CC1CC[C@](C)(O1)C1CC[C@](C)(O1)[C@@H](C)O | |
| 709.02 |
Safety and Handling
Recommended Storage : Store at -20°C