missing translation for 'onlineSavingsMsg'
Learn More

Ionic Strength Adjustor for Sodium Testing, Reagents

Catalog No. p-200021980
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334927 1 L
01334926 4 L
01334925 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334927 Supplier Labchem Inc Supplier No. CI1800001A
Only null left

Ionic Strength Adjustor for Sodium Testing, Reagent, use for Sodium analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 12125-02-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Poly Bottle
Specific Gravity 1.03
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Sodium Analysis
Quantity 1 L
Grade Reagent
Chemical Name or Material Ionic Strength Adjustor for Sodium Testing
Show More Show Less