missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ionic Strength Adjustor for Sodium Testing, Reagents
Specifications
Specifications
| Packaging | Poly Bottle |
| Specific Gravity | 1.03 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| For Use With (Equipment) | Water Analyzer |
| Solubility | Completely miscible |
| Industry Type | Sodium Analysis |
| Quantity | 1 L |
| Grade | Reagent |
| Show More |