missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate-S Standard for Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$328.56 - $575.19
Specifications
| Packaging | Triple-leached LDPE bottle |
|---|---|
| CAS Min % | 0.1,99.9 |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| CAS Max % | 0.1,99.9 |
| CAS | 7732-18-5 |
| Technique | Ion Chromatography |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Solution Type | Standard Solution |
| Type | Certified Reference Material |
| Solvent or Matrix | Water |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AS-SO4S9-2Y
|
SPEX CertiPrep Group LLC
ASSO4S92Y |
125 mL | Triple-leached LDPE bottle |
N/A
|
|
||||
|
AS-SO4S9-2X
|
SPEX CertiPrep Group LLC
ASSO4S92X |
500 mL | Triple-leached LDPE bottle |
N/A
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| SO4-S | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 7732-18-5 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Certified Reference Material | |
| 125 mL |
| 0.1,99.9 | |
| 0.1,99.9 | |
| Ion Chromatography | |
| Standard Solution | |
| Water | |
| Sulfate-Sulfur Standard |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only