missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate Eluent Concentrate For Ion Chromatography, 0.5M, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ICELCON2100
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| NaHCO3 | 0.5M |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Sodium Bicarbonate Eluent Concentrate | |
| With Certificate of Analysis | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00003528 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Certified Reference Material | |
| [Na+].OC([O-])=O | |
| sodium;hydrogen carbonate | |
| 125 mL | |
| CHEBI:32139 |
| Triple-leached LDPE bottle | |
| 144-55-8,7732-18-5 | |
| CHNaO3 | |
| Ion Chromatography | |
| Standard Solution | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| Water | |
| 84.01 | |
| 516892 |
Chemical Identifiers
| 144-55-8 | |
| 84.01 | |
| UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
| CHEBI:32139 | |
| [Na+].OC([O-])=O |
| CHNaO3 | |
| MFCD00003528 | |
| 516892 | |
| sodium;hydrogen carbonate |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only