missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoride Standard for Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$307.47 - $640.03
Chemical Identifiers
| CAS | 7732-18-5 |
|---|
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| F- | 10μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7732-18-5 |
Specifications
| Triple-leached LDPE bottle | |
| 7732-18-5,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| Ion Chromatography | |
| Standard Solution | |
| Water | |
| H20 |
| With Certificate of Analysis. | |
| 0.0,99.99 | |
| 0.0,99.99 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Certified Reference Material | |
| 125 mL | |
| Fluoride Standard |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only