Learn More
Thermo Scientific™ Ion 16S™ Metagenomics Kit
The Ion 16S™ Metagenomics Kit is designed for rapid, comprehensive and broad-range analyses of mixed microbial populations using the Ion Torrent™ semiconducter sequencing workflow.
Supplier: Thermo Scientific™ A26216
Description
The Ion 16S™ Metagenomics Kit is designed for rapid, comprehensive and broad-range analyses of mixed microbial populations using the Ion Torrent™ semiconducter sequencing workflow. The kit permits PCR amplification of hypervariable regions of the 16S rDNA gene from bacteria. The amplified fragments can then be sequenced using the Ion PGM™ Sequencing 400 Kit on the Ion PGM™ platform and analyzed using the Ion 16S™ metagenomics analyses module within the Ion Reporter™ software.
Comprehensive
The kit includes 2 sets of primers that can be used to amplify the corresponding hypervariable regions of the 16S rDNA gene in bacteria:
- Primer set V2-4-8
- Primer set V3-6,7-9
These comprehensive primer sets allow for accurate detection and identification of a broad range of bacteria down to the genus or species level. The primer sets are paired with Environmental Master Mix v2.0 that is optimized to tolerate high levels of PCR inhibitors and amplify targets from complex samples such as environmental, food, tissue and other challenging samples. Additionally, the kit includes a positive control for troubleshooting experiments.
Flexible
The primer sets are provided as two pools. Each pool can be used individually or in combination to allow for flexibility in experimental set up. When combined with the Ion Xpress™ Barcode Adapters kits, multiple polymicrobial samples can be combined in a single sequencing workflow.
Complete Workflow
Sequences generated using the Ion 16S™ Metagenomics Kit can be directly analyzed using the Ion 16S™ metagenomics analyses module within the Ion Reporter™ software enabling a rapid and semi-quantitative assessment of complex microbial samples.
Note: This kit is not compatible with the Ion Library Quantitation Kit (Cat. No. 4468802). For accurate quantitation of amplicons generated using this kit, use the Ion Universal Library Quantitation Kit (Cat. No. A26217).
Specifications
| • DNA dilution buffer (7 mL) • 2 tubes 2X environmental master mix (0.8 mL per tube) Store the above at 2-8°C. • E. coli DNA control (40 μL) • Negative control water (1 mL) • 16S primer set V2-4-8 (300 μL) • 16S primer set V3-6,7-9 (300 μL) Store the above at -5 to -30°C. |
|
| DNA | |
| Sequencing | |
| Taq DNA Polymerase | |
| 100 reactions | |
| All Bacteria |
| Ion Torrent Sequencing, Primer-Probe Detection | |
| Ion GeneStudio™ S5 Series Sequencer | |
| 250 Reactions | |
| Ion Torrent | |
| All Species | |
| Fast |
For Research Use Only. Not for use in diagnostic procedures.