missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iohexol

Catalog No. AC466650250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466650250 25 g
AC466651000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466650250 Supplier Thermo Scientific Chemicals Supplier No. 466650250

CAS: 66108-95-0 | C19H26I3N3O9, C19H26I3N3O9 | 821.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66108-95-0
Molecular Formula C19H26I3N3O9

Specifications

Quantity 25 g
Solubility Solubility aq. soln.: soluble.
Melting Point 180.0°C
Formula Weight 821.13
InChI Key NTHXOOBQLCIOLC-UHFFFAOYNA-N
SMILES CC(=O)N(CC(O)CO)C1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
IUPAC Name N1,N3-bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodobenzene-1,3-dicarboxamide
Molecular Weight (g/mol) 821.14
Chemical Name or Material Iohexol
EINECSNumber 266-164-2

RUO – Research Use Only