missing translation for 'onlineSavingsMsg'
Learn More

Iodopentafluorobenzene, 99%, stabilised over copper

Catalog No. p-4323614
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312200010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC312200010 Supplier Thermo Scientific Chemicals Supplier No. 312200010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 827-15-6 | C6F5I | 293.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 827-15-6 , 7440-50-8
Molecular Formula C6F5I
Molecular Weight (g/mol) 293.96
InChI Key OPYHNLNYCRZOGY-UHFFFAOYSA-N
Synonym iodopentafluorobenzene, pentafluoroiodobenzene, benzene, pentafluoroiodo, 2,3,4,5,6-pentafluoroiodobenzene, iodoperfluorobenzene, pentafluorophenyl iodide, 1,2,3,4,5-pentafluoro-6-iodo-benzene, perfluoroiodobenzene, ibf, pubchem19982
PubChem CID 70008
IUPAC Name 1,2,3,4,5-pentafluoro-6-iodobenzene
SMILES C1(=C(C(=C(C(=C1F)F)I)F)F)F

Specifications

Color Colorless
Density 2.2040g/mL
Boiling Point 162.0°C to 165.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.496
Quantity 1 g
Specific Gravity 2.204
Formula Weight 293.96
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Iodopentafluorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 212-565-2

RUO – Research Use Only