missing translation for 'onlineSavingsMsg'
Learn More

Iodobenzene Diacetate 97.0+%, TCI America™

Catalog No. I033010G
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I033010G 10 g
I033025G 25 g
I0330250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I033010G Supplier TCI America Supplier No. I033010G

Chemical Identifiers

CAS 3240-34-4
Molecular Formula C10H11IO4
Molecular Weight (g/mol) 322.098
MDL Number MFCD00008692
InChI Key ZBIKORITPGTTGI-UHFFFAOYSA-N
Synonym iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo
PubChem CID 76724
IUPAC Name [acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
SMILES CC(=O)OI(C1=CC=CC=C1)OC(=O)C

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 10 g
Formula Weight 322.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iodobenzene Diacetate
RTECSNumber DA3525000
TSCA Yes