accessibility menu, dialog, popup

UserName

CAS RN 3240-34-4

IUPAC Name: (acetyloxy)(phenyl)-λ³-iodanyl acetate
Synonyms: phenyliodosodiacetate
Molecular Weight (g/mol): 322.1
Molecular Formula: C10H11IO4
InChi Key: ZBIKORITPGTTGI-UHFFFAOYSA-N
SMILES: CC(=O)O[I](OC(C)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)