Learn More
Iodoacetyl chloride, 97%
CAS: 38020-81-4 | C2H2ClIO | 204.39 g/mol
Supplier: Thermo Scientific Chemicals L0442814
Description
Iodoacetyl chloride is used for biochemical research purposes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodoacetyl chloride is used for biochemical research purposes.
Solubility
Miscible with water.
Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents. Reacts with water.
Specifications
| Iodoacetyl chloride | |
| 2.28 | |
| C2H2ClIO | |
| 25 g | |
| 1739078 | |
| iodoacetyl chloride, iodoacetylchloride, acetyl chloride, iodo, jodacetylchlorid, acetylchloride,iodo, iodoacetic acid chloride, acmc-1aewu, iodoacetyl chloride 5g, acetyl chloride, iodo-6ci,9ci | |
| BSVMPWANOMFSPR-UHFFFAOYSA-N | |
| 2-iodoacetyl chloride | |
| 3084680 | |
| 97% |
| 38020-81-4 | |
| 48°C to 50°C (12 mmHg) | |
| MFCD00013718 | |
| UN3265 | |
| Light and moisture sensitive | |
| Miscible with water. | |
| ClC(=O)CI | |
| 204.39 | |
| 204.39 |
Chemical Identifiers
| 38020-81-4 | |
| 204.39 | |
| BSVMPWANOMFSPR-UHFFFAOYSA-N | |
| 3084680 | |
| ClC(=O)CI |
| C2H2ClIO | |
| MFCD00013718 | |
| iodoacetyl chloride, iodoacetylchloride, acetyl chloride, iodo, jodacetylchlorid, acetylchloride,iodo, iodoacetic acid chloride, acmc-1aewu, iodoacetyl chloride 5g, acetyl chloride, iodo-6ci,9ci | |
| 2-iodoacetyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 253-748-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only