missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodoacetyl chloride, 97%
Description
Iodoacetyl chloride is used for biochemical research purposes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iodoacetyl chloride is used for biochemical research purposes.
Solubility
Miscible with water.
Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents. Reacts with water.
Iodoacetyl chloride is used for biochemical research purposes.
Solubility
Miscible with water.
Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents. Reacts with water.
Specifications
Specifications
| Density | 2.28 |
| Boiling Point | 48°C to 50°C (12 mmHg) |
| Quantity | 5 g |
| UN Number | UN3265 |
| Beilstein | 1739078 |
| Sensitivity | Light and moisture sensitive |
| Solubility Information | Miscible with water. |
| Formula Weight | 204.39 |
| Percent Purity | 97% |
| Chemical Name or Material | Iodoacetyl chloride |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 253-748-7 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only