missing translation for 'onlineSavingsMsg'
Learn More

Iodine Solution, Wijs, Fisher Chemical™

Catalog No. p-213481
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SI106500 500 mL Amber Glass
SI1064 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SI106500 Supplier Thermo Fisher Scientific Supplier No. SI106 500
Only null left

For determination of iodine absorption number in fats. Approximately 0.22N solution in glacial acetic acid

Chemical Identifiers

CAS 7553-56-2 , 64-19-7 , 7782-50-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Color Amber
Boiling Point 118°C
Melting Point 17°C
UN Number 2920
Concentration 0.215 to 0.225
Density 1.06g/cm³
Identification Pass test
Packaging Amber Glass
Vapor Pressure 11mmHg at 20°C
pH 2.0
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Iodine Solution, Wijs
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Explosive. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:2
Instability:0
DOTInformation DOT Class 8, : Corrosive