missing translation for 'onlineSavingsMsg'
Learn More

Iodine solution, according to Wijs, 0.1 mol ICl/l

Catalog No. p-215398
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270570010 1 L Glass bottle
AC270570025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270570010 Supplier Thermo Scientific Chemicals Supplier No. 270570010
Only null left

CAS: 7790-99-0 | ClI | 162.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-99-0 , 64-19-7
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Color Brown
Boiling Point 117.8°C
Melting Point 17.2°C
Concentration 0.098 to 0.113M
Fieser 16,182
Density 1.0600g/mL
Merck Index 15, 5060
Formula Weight 162.36
Packaging Glass bottle
Specific Gravity 1.06
Quantity 1 L
Flash Point 40°C
Physical Form Liquid
Chemical Name or Material Iodine solution
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only