missing translation for 'onlineSavingsMsg'
Learn More

Iodine and Potassium Iodide TS 1, Ricca Chemical

$353.19 - $518.06

Specifications

Chemical Name or Material Iodine and Potassium Iodide
Color Brown
Name Note Test Solution
CAS 7732-18-5,7681-11-0,7553-56-2
CAS Min % 91.0,5.45,1.82
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
4124-16
View Documents
Ricca Chemical Company
412416
500 mL
Each for $353.19
Only null left
 
4124-32
View Documents
Ricca Chemical Company
412432
1 L
Each for $518.06
Only null left
 
Description

Description

Specifications

Specifications

Iodine and Potassium Iodide
Test Solution
91.0,5.45,1.82
dihydrogen oxide, dihydrogen monoxide
Amber Glass Bottle
Brown
7732-18-5,7681-11-0,7553-56-2
94.0,5.67,1.89
Laboratory
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'