Learn More
Iodine and Potassium Iodide TS 1, Ricca Chemical
Supplier: Ricca Chemical Company 412432
Specifications
| Iodine and Potassium Iodide | |
| Test Solution | |
| 91.0,5.45,1.82 | |
| dihydrogen oxide, dihydrogen monoxide | |
| Amber Glass Bottle | |
| Liquid |
| Brown | |
| 7732-18-5,7681-11-0,7553-56-2 | |
| 94.0,5.67,1.89 | |
| Laboratory | |
| 1 L |