missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride Solution, Wijs, for Iodine Absorption Number of Fats and Oils, Ricca Chemical

Catalog No. 411016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
411016 500 mL Amber Glass Bottle
411032 1 L Amber Glass Bottle
41101 4 L Amber Glass Bottle
13622603 4 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 411016 Supplier Ricca Chemical Company Supplier No. 411016

For Analysis of Iodine Absorption Number of Fats and Oils

Chemical Identifiers

CAS 64-19-7 , 7790-99-0 , 7553-56-2
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 96.0, 1.41, 0.09
CAS Max % 100.0, 1.46, 0.1
Grade Laboratory
Packaging Amber Glass Bottle
pH <1
Quantity 500 mL
Chemical Name or Material Iodine Monochloride Solution, Wijs