missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine (Iodine-Iodide), 0.0200 N (N/50), Ricca Chemical
$78.50 - $341.66
Chemical Identifiers
| Molecular Formula | I2 |
|---|---|
| MDL Number | MFCD00011355 MFCD00164163 |
| InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
| Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
| PubChem CID | 807 |
| ChEBI | CHEBI:17606 |
| IUPAC Name | diiodine |
| SMILES | II |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
397316
|
Ricca Chemical Company
397316 |
500 mL | Amber Glass Bottle |
Each for $78.50
|
|
||||
|
3973-32
|
Ricca Chemical Company
397332 |
1 L | Amber Glass Bottle |
Each for $99.56
|
|
||||
|
3973-1
|
Ricca Chemical Company
39731 |
4 L | Amber Glass Bottle |
Each for $341.66
|
|
||||
Chemical Identifiers
| I2 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| 807 | |
| diiodine |
| MFCD00011355 MFCD00164163 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| CHEBI:17606 | |
| II |
Specifications
| Brown | |
| 0.2,97.0,0.65 | |
| I2 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| II | |
| 253.81 | |
| CHEBI:17606 | |
| Amber Glass Bottle | |
| Liquid | |
| Iodine, Iodine-Iodide |
| 7553-56-2,7732-18-5,7681-11-0 | |
| 0.21,100.0,0.67 | |
| MFCD00011355 MFCD00164163 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| diiodine | |
| 807 | |
| Laboratory | |
| 500 mL | |
| 0.0200Normal |