missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine (Iodine-Iodide), 0.0200 N (N/50), Ricca Chemical
Supplier: Ricca Chemical Company 397316
Specifications
| Iodine | |
| Iodine-Iodide | |
| 0.2,97.0,0.65 | |
| I2 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| II | |
| 253.81 | |
| CHEBI:17606 | |
| Amber Glass Bottle | |
| Liquid |
| Brown | |
| 7553-56-2,7732-18-5,7681-11-0 | |
| 0.21,100.0,0.67 | |
| MFCD00011355 MFCD00164163 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| diiodine | |
| 807 | |
| Laboratory | |
| 500 mL | |
| 0.0200Normal |
Chemical Identifiers
| I2 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| 807 | |
| diiodine |
| MFCD00011355 MFCD00164163 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| CHEBI:17606 | |
| II |