missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, D'Antoni, Certified, 1.5% (w/v) ±0.1%, LabChem™
Specifications
Specifications
| Color | Brown-Red |
| CAS Max % | 1.5, 95.5, 3 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Linear Formula | I2 |
| Solubility Information | Soluble in water |
| Formula Weight | 253.809 |
| Percent Purity | 1.5% ±0.1% (w/v) |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Avoid breathing mist, vapors, spray. Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Contaminated work clothing must not be allowed out of the workplace. Avoid release to the environment. If on skin: Wash skin with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention. Take off contaminated clothing and wash it before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |